Merck
Haemodynamic and anatomic progression of aortic stenosis.
Heart (British Cardiac Society) (2015-02-07)
Virginia Nguyen, Claire Cimadevilla, Candice Estellat, Isabelle Codogno, Virginie Huart, Joelle Benessiano, Xavier Duval, Philippe Pibarot, Marie Annick Clavel, Maurice Enriquez-Sarano, Alec Vahanian, David Messika-Zeitoun
ABSTRACT
Aortic valve stenosis (AS) is a progressive disease, but the impact of baseline AS haemodynamic or anatomic severity on AS progression remains unclear. In 149 patients (104 mild AS, 36 moderate AS and 9 severe AS) enrolled in 2 ongoing prospective cohorts (COFRASA/GENERAC), we evaluated AS haemodynamic severity at baseline and yearly, thereafter, using echocardiography (mean pressure gradient (MPG)) and AS anatomic severity using CT (degree of aortic valve calcification (AVC)). After a mean follow-up of 2.9±1.0 years, mean MGP increased from 22±11 to 30±16 mm Hg (+3±3 mm Hg/year), and mean AVC from 1108±891 to 1640±1251 AU (arbitrary units) (+188±176 AU/year). Progression of AS was strongly related to baseline haemodynamic severity (+2±3 mm Hg/year in mild AS, +4±3 mm Hg/year in moderate AS and +5±5 mm Hg/year in severe AS (p=0.01)), and baseline haemodynamic severity was an independent predictor of haemodynamic progression (p=0.0003). Annualised haemodynamic and anatomic progression rates were significantly correlated (r=0.55, p<0.0001), but AVC progression rate was also significantly associated with baseline haemodynamic severity (+141±133 AU/year in mild AS, +279±189 AU/year in moderate AS and +361±293 AU/year in severe AS, p<0.0001), and both baseline MPG and baseline AVC were independent determinants of AVC progression (p<0.0001). AS progressed faster with increasing haemodynamic or anatomic severity. Our results suggest that a medical strategy aimed at preventing AVC progression may be useful in all subsets of patients with AS including those with severe AS and support the recommended closer follow-up of patients with AS as AS severity increases. COFRASA (clinicalTrial.gov number NCT 00338676) and GENERAC (clinicalTrial.gov number NCT00647088).
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis