Merck
  • A simple and rapid creatinine sensing via DLS selectivity, using calix[4]arene thiol functionalized gold nanoparticles.
A simple and rapid creatinine sensing via DLS selectivity, using calix[4]arene thiol functionalized gold nanoparticles.
Talanta (2015-11-26)
Pinkesh G Sutariya, Alok Pandya, Anand Lodha, Shobhana K Menon
ABSTRACT
A new, simple, ultra-sensitive and selective approach has been reported for the "on spot" colorimetric detection of creatinine based on calix[4]arene functionalized gold nanoparticles (AuNPs) with excellent discrimination in the presence of other biomolecules. The lower detection limit of the method is 2.16nM. The gold nanoparticles and p-tert-butylcalix[4]arene were synthesized by microwave assisted method. Specifically, in our study, we used dynamic light scattering (DLS) which is a powerful method for the determination of small changes in particle size, improved selectivity and sensitivity of the creatinine detection system over colorimetric method. The nanoassembly is characterized by Transmission electron microscopy (TEM), DLS, UV-vis and ESI-MS spectroscopy, which demonstrates the binding affinity due its ability of hydrogen bonding and electrostatic interaction between -NH group of creatinine and pSDSC4. It exhibits fast response time (<60s) to creatinine and has long shelf-life (>5 weeks). The developed pSDSC4-AuNPs based creatinine biosensor will be established as simple, reliable and accurate tool for the determination of creatinine in human urine samples.
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Pricing and availability is not currently available.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Pricing and availability is not currently available.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Pricing and availability is not currently available.
Supelco
L-Ascorbic acid, analytical standard
Pricing and availability is not currently available.
Supelco
Menaquinone (K2), analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Pricing and availability is not currently available.