Merck
  • Combining selection valve and mixing chamber for nanoflow gradient generation: Toward developing a liquid chromatography cartridge coupled with mass spectrometer for protein and peptide analysis.
Combining selection valve and mixing chamber for nanoflow gradient generation: Toward developing a liquid chromatography cartridge coupled with mass spectrometer for protein and peptide analysis.
Analytica chimica acta (2015-09-01)
Apeng Chen, Joann J Lu, Congying Gu, Min Zhang, Kyle B Lynch, Shaorong Liu
ABSTRACT
Toward developing a micro HPLC cartridge, we have recently built a high-pressure electroosmotic pump (EOP). However, we do not recommend people to use this pump to deliver an organic solvent directly, because it often makes the pump rate unstable. We have experimented several approaches to address this issue, but none of them are satisfactory. Here, we develop an innovative approach to address this issue. We first create an abruption (a dead-volume) within a fluid conduit. We then utilize an EOP to withdraw, via a selection valve, a train of eluent solutions having decreasing eluting power into the fluid conduit. When these solutions are further aspirated through the dead-volume, these solutions are partially mixed, smoothening concentration transitions between two adjacent eluent solutions. As these solutions are pushed back, through the dead-volume again, a smooth gradient profile is formed. In this work, we characterize this scheme for gradient formation, and we incorporate this approach with a high-pressure EOP, a nanoliter injection valve, and a capillary column, yielding a micro HPLC system. We then couple this micro HPLC with an electrospray ionization - mass spectrometer for peptide and protein separations and identifications.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Butyl methacrylate, 99%, contains monomethyl ether hydroquinone as inhibitor