Merck
  • A High Throughput Method for Estimating Mouth-Level Intake of Mainstream Cigarette Smoke.
A High Throughput Method for Estimating Mouth-Level Intake of Mainstream Cigarette Smoke.
Nicotine & tobacco research : official journal of the Society for Research on Nicotine and Tobacco (2015-02-05)
Xizheng Yan, Liqin Zhang, Bryan A Hearn, Liza Valentín-Blasini, Gregory M Polzin, Clifford H Watson
ABSTRACT
We developed a high throughput method for estimating smoker's mainstream smoke intake on a per-cigarette basis by analyzing discarded cigarette butts. This new method utilizes ultraviolet/visible (UV-Vis) spectrophotometric analysis of isopropanol-soluble smoke particulate matter extracted from discarded cigarette filters. When measured under a wide range of smoking conditions for a given brand variant, smoking machine delivery of nicotine, benzene, polycyclic aromatic hydrocarbons, and tobacco-specific nitrosamines can be related to the overall filter extract absorbance at 360 nm. Once this relationship has been established, UV-Vis analysis of a discarded cigarette filter butt gives a quantitative measure of a smoker's exposure to these analytes. The measured mainstream smoke constituents correlated closely (correlation coefficients from 0.9303 to 0.9941) with the filter extract absorbance. These high correlations held over a wide range of smoking conditions for 2R4F research cigarettes as well as popular domestic cigarette brands sold in the United States. This low cost, high throughput method is suitable for high volume analyses (hundreds of samples per day) because UV-Vis spectrophotometry, rather than mass spectrometry, is used for the cigarette filter butt analysis. This method provides a stable and noninvasive means for estimating mouth-level delivery of many mainstream smoke constituents. The ability to gauge the mouth-level intake of harmful chemicals and total mainstream smoke for cigarette smokers in a natural setting on a cigarette-by-cigarette basis can provide insights on factors contributing to morbidity and mortality from cigarette smoking, as well as insights on strategies related to smoking cessation.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%