Merck
  • The metabolism of histamine in rat hypothalamus and cortex after reserpine treatment.
The metabolism of histamine in rat hypothalamus and cortex after reserpine treatment.
Neurochemistry international (2015-05-06)
Martin Maldonado, Kazutaka Maeyama
ABSTRACT
The effect of reserpine on histamine (HA) and tele-methylhistamine (N(τ)-MHA) in hypothalamus and cortex of rats was analyzed and compared to catecholamines. IP injection of reserpine (5 mg/kg) confirmed the effectiveness of reserpine treatment on noradrenaline and dopamine levels. Our in-vitro experiment with synaptosomal/crude mitochondrial fraction from hypothalamus and cortex confirmed that while mono amine oxidase (MAO) is an efficient metabolic enzyme for catecholamines, HA is not significantly affected by its enzymatic action. HMT activity after reserpine, pargyline and L-histidine treatment showed no differences compared to the control values. However HDC was significantly increased in both hypothalamus and cortex. In this study, Ws/Ws rats with deficiency of mast cells were used to clarify aspects of HA metabolism in HAergic neurons by eliminating the contribution of mast cells. The irreversible MAO-B inhibitor Pargyline (65 mg/kg) failed to accumulate N(τ)-MHA in the hypothalamus. However, when animals treated with reserpine and pargyline/reserpine were compared, the last group showed higher N(τ)-MHA values (p < 0.01). Moreover, the precursor of HA, L-histidine (1 g/kg), produced an increase of HA in the hypothalamus to 166% and the cortex to 348%. In conclusion, our results suggest that the effect of reserpine on the HA pools in the brain might be different. The neuronal HA pools are more resistant to reserpine as compared to those of catecholamine. Moreover, the HAergic pool appears to be more resistant to depletion than mast cells' pool, and thus HDC/HMT activity and its localization may play a key role in the understanding of HA metabolism in brain after reserpine treatment.
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
SAFC
L-Histidine monohydrochloride monohydrate
Sigma-Aldrich
L-Histidine monohydrochloride monohydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine monohydrochloride monohydrate, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Phthaldialdehyde, ≥99.0% (HPLC)
Sigma-Aldrich
L-Histidine monohydrochloride monohydrate, ≥99.0% (AT)
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Pargyline hydrochloride
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%