Merck
  • Novel Coumarin Substituted Water Soluble Cyclophosphazenes as "Turn-Off" Type Fluorescence Chemosensors for Detection of Fe(3+) ions in Aqueous Media.
Novel Coumarin Substituted Water Soluble Cyclophosphazenes as "Turn-Off" Type Fluorescence Chemosensors for Detection of Fe(3+) ions in Aqueous Media.
Journal of fluorescence (2015-10-01)
Gönül Yenilmez Çiftçi, Elif Şenkuytu, Mustafa Bulut, Mahmut Durmuş
ABSTRACT
In the present work, 3-[2-(diethylamino)ethyl]-7-oxy-4-methylcoumarin substituted cyclotriphosphazene (4) and cyclotetraphosphazene (5) derivatives were synthesized by the reactions of hexachlorocyclotriphosphazene (1) or octachlorocyclotetraphosphazene (2) with 3-[2-(diethylamino)ethyl]-7-hydroxy-4-methylcoumarin (3) for the first time. The quaternized cationic (6 and 7) and zwitterionic (8 and 9) derivatives of these compounds (4 and 5) were obtained by the reactions of dimethyl sulfate and 1,3-propanesultone, respectively. All newly synthesized cyclophosphazene compounds (4-9) were fully characterized by elemental analysis and general spectroscopic techniques such as FT-IR, (31)P-NMR, (1)H-NMR and MALDI-TOF mass. All these coumarin substituted cyclophosphazene compounds (4-9) were soluble in most of organic solvents and quaternized ionic and zwitterionic compounds (6-9) also showed excellent solubility in water. The fluorescence behaviors of novel cyclophosphazene compounds were investigated in methanol and water solutions. The chemosensor properties of newly synthesized water soluble quaternized ionic and zwitterionic cyclotriphosphazene and cyclotetraphosphazene derivatives (6-9) were investigated in aqueous media. These cyclophosphazene derivatives showed fluorescence chemosensor behavior with high selectivity for Fe(3+) ions in aqueous solution.
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Cesium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
1,3-Propanesultone, ≥99%
Sigma-Aldrich
1,3-Propanesultone, 98%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Cesium carbonate, 99.95% trace metals basis
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D