Merck
  • CRL3IBTK Regulates the Tumor Suppressor Pdcd4 through Ubiquitylation Coupled to Proteasomal Degradation.
CRL3IBTK Regulates the Tumor Suppressor Pdcd4 through Ubiquitylation Coupled to Proteasomal Degradation.
The Journal of biological chemistry (2015-04-18)
Antonio Pisano, Simona Ceglia, Camillo Palmieri, Eleonora Vecchio, Giuseppe Fiume, Annamaria de Laurentiis, Selena Mimmi, Cristina Falcone, Enrico Iaccino, Annarita Scialdone, Marilena Pontoriero, Francesca Fasanella Masci, Rosanna Valea, Shibu Krishnan, Marco Gaspari, Giovanni Cuda, Giuseppe Scala, Ileana Quinto
ABSTRACT
The human inhibitor of Bruton's tyrosine kinase isoform α (IBtkα) is a BTB protein encoded by the IBTK gene, which maps to chromosomal locus 6q14.1, a mutational hot spot in lymphoproliferative disorders. Here, we demonstrate that IBtkα forms a CRL3(IBTK) complex promoting its self-ubiquitylation. We identified the tumor suppressor Pdcd4 as IBtkα interactor and ubiquitylation substrate of CRL3(IBTK) for proteasomal degradation. Serum-induced degradation of Pdcd4 required both IBtkα and Cul3, indicating that CRL3(IBTK) regulated the Pdcd4 stability in serum signaling. By promoting Pdcd4 degradation, IBtkα counteracted the suppressive effect of Pdcd4 on translation of reporter luciferase mRNAs with stem-loop structured or unstructured 5'-UTR. IBtkα depletion by RNAi caused Pdcd4 accumulation and decreased the translation of Bcl-xL mRNA, a well known target of Pdcd4 repression. By characterizing CRL3(IBTK) as a novel ubiquitin ligase, this study provides new insights into regulatory mechanisms of cellular pathways, such as the Pdcd4-dependent translation of mRNAs.
Product Description
SAFC
L-Glutamine
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atp8a2
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP8A2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cul3
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CUL3
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)