Merck
  • Quantitative aspects of electrolysis in electromembrane extractions of acidic and basic analytes.
Quantitative aspects of electrolysis in electromembrane extractions of acidic and basic analytes.
Analytica chimica acta (2015-09-01)
Andrea Šlampová, Pavel Kubáň, Petr Boček
ABSTRACT
Electrolysis is omnipresent in all electrochemical processes including electromembrane extraction (EME). The effects of electrolysis on quantitative aspects of EME were comprehensively evaluated for a set of acidic (substituted phenols) and basic (basic drugs) analytes. EMEs were carried out across supported liquid membranes formed by 1-ethyl-2-nitrobenzene at standard EME conditions, i.e., acidic analytes were extracted from alkaline into alkaline solutions and basic analytes were extracted from acidic into acidic solutions. Electric potential applied across the EME systems was 50 V and extraction recoveries of analytes as well as pH values of donor and acceptor solutions were determined after each EME. It has been proven that electrolysis plays a more significant role than has ever been thought before in EME. Electrolytically produced H(+) and OH(-) ions had a significant effect on pH values of acceptor solutions and variations of up to 8.5 pH units were obtained at standard EME conditions. pH values of donor solutions were affected only negligibly due to their significantly higher volumes. The observed variations in pH values of acceptor solutions had fatal consequences on quantitative EME results of weak and medium strong acidic/basic analytes. A direct relation was observed between the decrease in extraction recoveries of the analytes, their pKa values and the acceptor solution pH values. Acceptor solutions consisting of high concentrations of weak bases or acids were thus proposed as suitable EME operational solutions since they efficiently eliminated the electrolytically induced pH variations, offered stable EME performances and were easily compatible with subsequent analytical methods.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
4-Chloro-DL-phenylalanine
Sigma-Aldrich
Cesium hydroxide hydrate, ≥90%, ≥99.5% (metals basis)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Cesium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Ethyl-2-nitrobenzene, 96%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Papaverine hydrochloride, powder
Sigma-Aldrich
Procaine hydrochloride, ≥97%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Cesium hydroxide monohydrate, ≥99.5% trace metals basis