Merck
  • Characterization of a highly sensitive and selective novel trapping reagent, stable isotope labeled glutathione ethyl ester, for the detection of reactive metabolites.
Characterization of a highly sensitive and selective novel trapping reagent, stable isotope labeled glutathione ethyl ester, for the detection of reactive metabolites.
Journal of pharmacological and toxicological methods (2015-09-01)
Toshikazu Yamaoka, Yoshiaki Kitamura
ABSTRACT
Glutathione (GSH) trapping assays are widely used to predict the post-marketing risk for idiosyncratic drug reactions (IDRs) in the pharmaceutical industry. Although several GSH derivatives have been introduced as trapping reagents for reactive intermediates, more sensitive and selective reagents are desired to prevent the generation of erroneous results. In this study, stable isotope labeled GSH ethyl ester (GSHEE-d5) was designed and its detection capability was evaluated. GSHEE-d5 was synthesized and its detection potential was compared with stable isotope labeled GSH ([(13)C2,(15)N]GSH) as a reference trapping reagent. The trapping reagents were added to human liver microsomes as a 1:1 mixture with GSHEE or GSH, respectively, and incubated with seven IDR positive drugs and three IDR negative drugs. The adducts formed between the reagents and reactive metabolites were analyzed by unit resolution mass spectrometer (MS) using isotope pattern-dependent scan with neutral loss filtering. A single-step reaction of GSH and ethanol-d6 produced GSHEE-d5 with a yield of 85%. The GSHEE-d5 assay detected adducts with all seven IDR positive drugs, and no adducts were detected with the three IDR negative drugs. In contrast, the [(13)C2,(15)N]GSH assay failed to detect adducts with three of the IDR positive drugs. In the case of diclofenac, the GSHEE-d5 assay showed a 4-times greater signal intensity than the [(13)C2,(15)N]GSH assay. GSHEE-d5 enabled the detection of reactive metabolites with greater sensitivity and selectivity than [(13)C2,(15)N]GSH. These results demonstrate that GSHEE-d5 would be a useful trapping reagent for evaluating the risk of IDRs with unit resolution MS.
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Mefenamic acid
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS