Merck
  • Genotype-driven isolation of enterocin with novel bioactivities from mangrove-derived Streptomyces qinglanensis 172205.
Genotype-driven isolation of enterocin with novel bioactivities from mangrove-derived Streptomyces qinglanensis 172205.
Applied microbiology and biotechnology (2015-04-22)
Dong-Bo Xu, Min Ma, Zi-Xin Deng, Kui Hong
ABSTRACT
The type II polyketide synthase (PKS) natural product enterocin (1) was isolated from a mangrove-derived novel species Streptomyces qinglanensis 172205 guided by genome sequence, and its putative biosynthetic gene cluster was revealed. Its natural analogues 5-deoxyenterocin (2) and wailupemycin A-C (3-5) were also identified by tandem mass spectrometry. By feeding experiments with aryl acids, strain 172205 was proved to incorporate partial exogenous starter units into enterocin- and wailupemycin-based analogues, thus being a new and suitable microorganism for engineering unnatural enc-derived polyketide metabolites. In addition, biological assays indicated that enterocin showed obvious inhibitory activity against β-amyloid protein (Aβ1-42) fibrillation and moderate cytotoxicity against HeLa and HepG2 for the first time.
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥99.0% (TLC)
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥98% (TLC)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Enterocin, ≥95% (HPLC)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Amyloid β Protein Fragment 1-42