Merck
  • Vimentin filament organization and stress sensing depend on its single cysteine residue and zinc binding.
Vimentin filament organization and stress sensing depend on its single cysteine residue and zinc binding.
Nature communications (2015-06-03)
Dolores Pérez-Sala, Clara L Oeste, Alma E Martínez, M Jesús Carrasco, Beatriz Garzón, F Javier Cañada
ABSTRACT
The vimentin filament network plays a key role in cell architecture and signalling, as well as in epithelial-mesenchymal transition. Vimentin C328 is targeted by various oxidative modifications, but its role in vimentin organization is not known. Here we show that C328 is essential for vimentin network reorganization in response to oxidants and electrophiles, and is required for optimal vimentin performance in network expansion, lysosomal distribution and aggresome formation. C328 may fulfil these roles through interaction with zinc. In vitro, micromolar zinc protects vimentin from iodoacetamide modification and elicits vimentin polymerization into optically detectable structures; in cells, zinc closely associates with vimentin and its depletion causes reversible filament disassembly. Finally, zinc transport-deficient human fibroblasts show increased vimentin solubility and susceptibility to disruption, which are restored by zinc supplementation. These results unveil a critical role of C328 in vimentin organization and open new perspectives for the regulation of intermediate filaments by zinc.
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Prostaglandin A1, synthetic
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diamide
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fast Blue B Salt, Dye content ~95 %
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Zinquin, ≥95% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Zinquin ethyl ester, ≥95% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%