Merck
  • Evaluation of less invasive methods to assess fatty acids from phospholipid fraction: cheek cell and capillary blood sampling.
Evaluation of less invasive methods to assess fatty acids from phospholipid fraction: cheek cell and capillary blood sampling.
International journal of food sciences and nutrition (2015-10-31)
Andrea de la Garza Puentes, Rosa Montes Goyanes, Aida Maribel Chisaguano Tonato, Ana Isabel Castellote, Rosario Moreno-Torres, Cristina Campoy Folgoso, M Carmen López-Sabater
ABSTRACT
Plasma is the most commonly employed matrix for analyzing fatty acids (FAs), but its extraction is not well accepted in the infant population. The objectives of this study were to evaluate cheek cells and capillary blood as alternatives to plasma sampling for FA analysis and to standardize the methodology. Samples were obtained from 20 children who underwent lipid extraction, phospholipid isolation by Solid Phase Extraction (SPE) in a 96-well plate, methylation, and analysis by fast gas chromatography (GC). A positive correlation was found for most of the FAs, especially long-chain polyunsaturated fatty acids (LC-PUFAs), in cheek cells and capillary blood versus plasma samples (r = 0.32-0.99). No differences were found in the levels of n-6: n-3 PUFA and n-6: n-3 LC-PUFA ratios between cheek cells and capillary blood. These two proposed samples can therefore be used as alternatives to plasma sampling for phospholipid FA analysis, especially LC-PUFAs.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Boron trifluoride-methanol solution, 14% in methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Boron trifluoride methanol complex solution, 13-15% BF3 basis
Sigma-Aldrich
Boron trifluoride-methanol solution, 50% w/w in methanol
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Sodium methoxide solution, 25 wt. % in methanol
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder