Merck
  • Galactosylated magnetic nanovectors for regulation of lipid metabolism based on biomarker-specific RNAi and MR imaging.
Galactosylated magnetic nanovectors for regulation of lipid metabolism based on biomarker-specific RNAi and MR imaging.
Nanotechnology (2015-08-01)
Dan Heo, Chanjoo Lee, Minhee Ku, Seungjoo Haam, Jin-Suck Suh, Yong-Min Huh, Sahng Wook Park, Jaemoon Yang
ABSTRACT
The specific delivery of ribonucleic acid (RNA) interfering molecules to disease-related cells is still a critical blockade for in vivo systemic treatment. Here, this study suggests a robust delivery carrier for targeted delivery of RNA-interfering molecules using galactosylated magnetic nanovectors (gMNVs). gMNVs are an organic-inorganic polymeric nanomaterial composed of polycationics and magnetic nanocrystal for delivery of RNA-interfering molecules and tracking via magnetic resonance (MR) imaging. In particular, the surface of gMNVs was modified by galactosylgluconic groups for targeted delivering to asialoglycoprotein receptor (ASGPR) of hepatocytes. Moreover, the small interfering RNAs were used to regulate target proteins related with low-density lipoprotein level and in vivo MR imaging was conducted for tracking of nanovectors. The obtained results show that the prepared gMNVs demonstrate potential as a systemic theragnostic nanoplatform for RNA interference and MR imaging.
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcsk9
Sigma-Aldrich
Lactobionic acid, ≥97% (TLC), cell impermeant agent
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
Lactobionic acid, 97% (TLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human PCSK9