Merck
  • Expression of voltage-gated calcium channel α(2)δ(4) subunits in the mouse and rat retina.
Expression of voltage-gated calcium channel α(2)δ(4) subunits in the mouse and rat retina.
The Journal of comparative neurology (2013-01-09)
Luis Pérez De Sevilla Müller, Janelle Liu, Alexander Solomon, Allen Rodriguez, Nicholas C Brecha
ABSTRACT
High-voltage activated Ca channels participate in multiple cellular functions, including transmitter release, excitation, and gene transcription. Ca channels are heteromeric proteins consisting of a pore-forming α(1) subunit and auxiliary α(2)δ and β subunits. Although there are reports of α(2)δ(4) subunit mRNA in the mouse retina and localization of the α(2)δ(4) subunit immunoreactivity to salamander photoreceptor terminals, there is a limited overall understanding of its expression and localization in the retina. α(2)δ(4) subunit expression and distribution in the mouse and rat retina were evaluated by using reverse transcriptase polymerase chain reaction, western blot, and immunohistochemistry with specific primers and a well-characterized antibody to the α(2)δ(4) subunit. α(2)δ(4) subunit mRNA and protein are present in mouse and rat retina, brain, and liver homogenates. Immunostaining for the α(2)δ(4) subunit is mainly localized to Müller cell processes and endfeet, photoreceptor terminals, and photoreceptor outer segments. This subunit is also expressed in a few displaced ganglion cells and bipolar cell dendrites. These findings suggest that the α(2)δ(4) subunit participates in the modulation of L-type Ca(2+) current regulating neurotransmitter release from photoreceptor terminals and Ca(2+)-dependent signaling pathways in bipolar and Müller cells.
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-G Protein Goα Antibody, clone 2A, clone 2A, Chemicon®, from mouse
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Decarboxylase Antibody, 65 kDa isoform, clone GAD-6, clone GAD-6, Chemicon®, from mouse