Merck
  • Long-term primary culture of a clear cell ovarian carcinoma reveals an epithelial-mesenchymal cooperative interaction.
Long-term primary culture of a clear cell ovarian carcinoma reveals an epithelial-mesenchymal cooperative interaction.
Cancer cell international (2015-09-26)
Alicia A Goyeneche, Michael Koch, Maria C Bell, Carlos M Telleria
ABSTRACT
We studied a primary culture developed from a biopsy of a clear cell carcinoma of the ovary (O-CCC) by (a) assessing its capacity to retain in vitro pathological features of the tumor of origin; (b) characterizing the main cells released from the complex mass without forced purification of any particular cellular entity; and (c) investigating its long-term proliferative capacity. A primary cell culture was developed from a pelvic mass diagnosed as an O-CCC. The morphological analysis of the cell culture was carried out by phase contrast microscopy. Markers of epithelial, mesenchymal, and tumor initiating cells were evaluated by immunocytochemistry. Cell proliferation was studied by detection of bromodeoxyuridine (BrdU) incorporated into newly synthesized DNA. As a biomarker of O-CCC, we assessed the expression of hepatocyte nuclear factor (HNF) 1β. We show that cells with epithelial morphological features express E-cadherin and expand with time in culture, a fact that the incorporation of BrdU confirms. Cells with mesenchymal-like characteristics that express the mesenchymal marker vimentin, however, allocate to the edges of the epithelial compartment. Moreover, we found that some cells with epithelial features also expressed vimentin. At the beginning of incubation, over 60 % of primary cells expressed the O-CCC marker HNF1β; such percentage declined upon passaging. We show that epithelial not mesenchymal cells undergo DNA replication, and that few cells in both epithelial and mesenchymal compartments express the stem-like tumor antigen CD133. We provide proof-of-principle that cells separated in bulk from a biopsy of an O-CCC can be maintained in culture for several months, and that two consistent cellular compartments-one epithelial that retains the O-CCC marker HNF1β, and another mesenchymal-persist, and seem to have a cooperative interaction leading to the multiplication of epithelial cells within a mesenchymal cellular environment.