Merck
  • Downregulation of the cancer susceptibility protein WRAP53β in epithelial ovarian cancer leads to defective DNA repair and poor clinical outcome.
Downregulation of the cancer susceptibility protein WRAP53β in epithelial ovarian cancer leads to defective DNA repair and poor clinical outcome.
Cell death & disease (2015-10-02)
E Hedström, C Pederiva, J Farnebo, B Nodin, K Jirström, D J Brennan, M Farnebo
ABSTRACT
Alterations in the scaffold protein WRAP53β have previously been linked to carcinogenesis and, in particular, associated with an increased risk for epithelial ovarian cancer. Here, we investigated the pathogenic impact and prognostic significance of WRAP53β in connection with epithelial ovarian cancer and examined the underlying mechanisms. We find that reduced expression of WRAP53β in ovarian tumors correlated with attenuated DNA damage response and poor patient survival. Furthermore, in ovarian cancer cell lines, WRAP53β was rapidly recruited to DNA double-strand breaks, where it orchestrated the recruitment of repair factors involved in homologous recombination and non-homologous end joining, including RNF168, 53BP1, BRCA1 and RAD51. Mechanistically, WRAP53β accomplishes this by facilitating the necessary ubiquitinylation at DNA breaks. Finally, we demonstrate that loss of WRAP53β significantly impairs the repair of DNA double-strand breaks, resulting in their accumulation. Our findings establish WRAP53β as a regulator of homologous recombination and non-homologous end joining repair in ovarian cancer cells, suggesting that loss of this protein contributes to the development and/or progression of ovarian tumors. Moreover, our current observations identify the nuclear levels of WRAP53β as a promising biomarker for the survival of patients with ovarian cancer.
Sigma-Aldrich
Anti-Ubiquitin Mouse mAb (FK2), liquid, clone FK2, Calbiochem®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-RNF168 Antibody, from rabbit, purified by affinity chromatography