Merck
The RNA binding protein quaking regulates formation of circRNAs.
Cell (2015-03-15)
Simon J Conn, Katherine A Pillman, John Toubia, Vanessa M Conn, Marika Salmanidis, Caroline A Phillips, Suraya Roslan, Andreas W Schreiber, Philip A Gregory, Gregory J Goodall
ABSTRACT
Circular RNAs (circRNAs), formed by non-sequential back-splicing of pre-mRNA transcripts, are a widespread form of non-coding RNA in animal cells. However, it is unclear whether the majority of circRNAs represent splicing by-products without function or are produced in a regulated manner to carry out specific cellular functions. We show that hundreds of circRNAs are regulated during human epithelial-mesenchymal transition (EMT) and find that the production of over one-third of abundant circRNAs is dynamically regulated by the alternative splicing factor, Quaking (QKI), which itself is regulated during EMT. Furthermore, by modulating QKI levels, we show the effect on circRNA abundance is dependent on intronic QKI binding motifs. Critically, the addition of QKI motifs is sufficient to induce de novo circRNA formation from transcripts that are normally linearly spliced. These findings demonstrate circRNAs are both purposefully synthesized and regulated by cell-type specific mechanisms, suggesting they play specific biological roles in EMT.
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Target shRNA Control Plasmid DNA, Targets no known genes from any species
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)