Merck
  • Promotion of regulatory T cell induction by immunomodulatory herbal medicine licorice and its two constituents.
Promotion of regulatory T cell induction by immunomodulatory herbal medicine licorice and its two constituents.
Scientific reports (2015-09-16)
Ao Guo, Dongming He, Hong-Bo Xu, Chang-An Geng, Jian Zhao
ABSTRACT
Regulatory T cells (Treg) play a critical role to control immune responses and to prevent autoimmunity, thus selective increase of Treg cells in vivo has broad therapeutic implications for autoimmune and inflammatory diseases. Licorice is a well-known herbal medicine used worldwide for over thousands of years, and accumulating evidence has shown its immunomodulatory potential. However, it is not clear whether licorice could regulate the induction and function of Treg cells. Here we found licorice extract could promote Treg cell induction, and then we used a rational approach to isolate its functional fractions and constituents. The results showed that two constituents, isoliquiritigenin and naringenin, promoted Treg cell induction both in vitro and in vivo. The effective fractions and two constituents of licorice also enhanced immune suppression of Treg cells, and they further reduced severity of DSS-induced colitis in mice. This study suggested that promotion of regulatory T cell induction could be an underlying mechanism of the historically and widely used herbal medicine licorice, providing its two effective molecules against autoimmune and inflammatory diseases.
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
JumpStart Taq ReadyMix for Quantitative PCR, For probe-based real-time PCR
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO