Merck
  • Characterization of a novel process-related impurity in commercial bendazac lysine eye drops by LC-ESI-QTOF/MS/MS and NMR.
Characterization of a novel process-related impurity in commercial bendazac lysine eye drops by LC-ESI-QTOF/MS/MS and NMR.
Journal of pharmaceutical and biomedical analysis (2015-02-11)
Peixi Zhu, Weifeng Yang, Liya Hong, Qiaoyuan Cheng, Lei Luo, Jiajia He, Juanjuan Chen
ABSTRACT
A simple and sensitive method was developed for characterization of two impurities from commercial bendazac lysine eye drops, by using liquid chromatography combined with electrospray ionization and QTOF mass analyzer (LC-ESI-QTOF MS/MS). A known impurity was characterized rapidly by comparison with the retention time and MS/MS data of the reference standard. Based on high resolution mass spectrometry (HRMS) and the nuclear magnetic resonance (NMR), a new impurity, purified by preparative HPLC, was identified as 2-(1,5-dibenzyl-1H-indazol-3-yloxy)acetic acid. In addition, a plausible mechanism for the formation of the impurities was also proposed.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%