Merck
  • Effect of clozapine and olanzapine on neutrophil kinetics: implications for drug-induced agranulocytosis.
Effect of clozapine and olanzapine on neutrophil kinetics: implications for drug-induced agranulocytosis.
Chemical research in toxicology (2014-06-27)
Winnie Ng, Rossoune Kennar, Jack Uetrecht
ABSTRACT
Clozapine is effective in the treatment of schizophrenia; however, its use is limited by a relatively high incidence of idiosyncratic agranulocytosis. The mechanism of clozapine-induced idiosyncratic agranulocytosis is unknown. Although most patients treated with clozapine do not develop agranulocytosis, most do have an immune response with an increase in inflammatory cytokines such as IL-6 and a release of immature neutrophils with neutrophilia rather than agranulocytosis. We have previously shown that treatment of rabbits with clozapine also causes an early release of neutrophils. Clozapine is oxidized to a reactive nitrenium ion that covalently binds to neutrophils, and this reactive metabolite may be responsible for the observed effects. Olanzapine and clozapine have very similar structures, and olanzapine is also oxidized to a reactive nitrenium ion; however, if it ever causes agranulocytosis, the incidence is much lower than that of clozapine. One possible basis for the difference in incidence of agranulocytosis between clozapine and olanzapine is that the therapeutic dose of olanzapine is much lower than that of clozapine. In this study, we compared the effects of clozapine and olanzapine in Sprague-Dawley rats at an equimolar dose and found that only clozapine had a significant effect on neutrophil kinetics. This suggests that the immune response and effects on neutrophil kinetics induced by clozapine are related to its ability to cause agranulocytosis.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, ≥98.0% (T), pellets
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Clozapine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %