Merck
  • Effect of hydrodynamics-based delivery of IL-18BP fusion gene on rat experimental autoimmune myocarditis.
Effect of hydrodynamics-based delivery of IL-18BP fusion gene on rat experimental autoimmune myocarditis.
Clinical and experimental medicine (2013-10-15)
He Chang, Yan Wang, Gang Li, Le Zhang, Guang Wei Zhang, Yan Chun Liao, Haruo Hanawa, Jun Zou
ABSTRACT
Interleukin-18 (IL-18) is a powerful and important cytokine in myocarditis. IL-18-binding protein (IL-18BP), a naturally occurring antagonist of IL-18, is presumed to play a vital regulatory function in IL-18-mediated immune responses. The purpose of this study was to evaluate the alterations of IL-18 and its related protein expressions and the effect of hydrodynamics-based delivery of the IL-18BP gene for treatment of rat experimental autoimmune myocarditis (EAM).Rats were immunized on Day 0 and killed on 2, 3 and 4 weeks to determine IL-18 and its related protein expression and target cells in EAM hearts. On Day 6, rats were injected with a recombinant plasmid encoding IL-18BP-Ig or SP-Ig. On Day 17, rats were detected with echocardiography and then be killed. IL-18BP gene therapy was effective in controlling EAM, as monitored by a decreased ratio of heart weight to body weight, reduced myocarditis areas, reduced expression of atrial natriuretic peptide, brain natriuretic peptide, IL-17, IFN-γ, IL-6 and IL-10. Furthermore, the effect of serum containing IL-18BP on the expression of immune-relevant genes in IL-1α-stimulated NC cells and splenocytes cultured from EAM rats was examined. The results showed that IL-18BP significantly suppressed the expression of IL-17 as well as other proinflammatory genes such as transforming growth factor-β, prostaglandin E2 synthase, cyclooxygenase-2 in IL-1α-stimulated NC cells, and IL-18BP also significantly suppressed the expression of IL-17, IL-17R, IL-21 and IL-17-related transcriptional factor retinoic acid-related orphan nuclear receptor, signal transducer and activator of transcription-3 and Foxp3 in IL-1α-stimulated splenocytes cultured from EAM rats. IL-18 and its related protein played an important role on the development of EAM. IL-18BP effectively prevented progression of EAM by blocking IL-17 and related inflammatory genes expression. This might be a possible mechanism of the amelioration of EAM by IL-18BP treatment.