Merck
  • The combination of electric current and copper promotes neuronal differentiation of adipose-derived stem cells.
The combination of electric current and copper promotes neuronal differentiation of adipose-derived stem cells.
Annals of biomedical engineering (2014-10-08)
L Jaatinen, S Salemi, S Miettinen, J Hyttinen, D Eberli
ABSTRACT
Damage to the nervous system can be caused by several types of insults, and it always has a great effect on the life of an individual. Due to the limited availability of neural transplants, alternative approaches for neural regeneration must be developed. Stem cells have a great potential to support neuronal regeneration. Human adipose-derived stem cells (hADSCs) have gained increasing interest in the fields of regenerative medicine due to their multilineage potential and easy harvest compared to other stem cells. In this study, we present a growth factor-free method for the differentiation of hADSCs toward neuron-like cells. We investigated the effect of electric current and copper on neuronal differentiation. We analyzed the morphological changes, the mRNA and protein expression levels in the stimulated cells and showed that the combination of current and copper induces stem cell differentiation toward the neuronal lineage with elongation of the cells and the upregulation of neuron-specific genes and proteins. The induction of the neuronal differentiation of hADSCs by electric field and copper may offer a novel approach for stem cell differentiation and may be a useful tool for safe stem cell-based therapeutic applications.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)