Merck
  • Promoter hypermethylation-related reduced somatostatin production promotes uncontrolled cell proliferation in colorectal cancer.
Promoter hypermethylation-related reduced somatostatin production promotes uncontrolled cell proliferation in colorectal cancer.
PloS one (2015-02-28)
Katalin Leiszter, Ferenc Sipos, Orsolya Galamb, Tibor Krenács, Gábor Veres, Barna Wichmann, István Fűri, Alexandra Kalmár, Árpád V Patai, Kinga Tóth, Gábor Valcz, Zsolt Tulassay, Béla Molnár
ABSTRACT
Somatostatin (SST) has anti-proliferative and pro-apoptotic effects. Our aims were to analyze and compare the SST expression during normal aging and colorectal carcinogenesis at mRNA and protein levels. Furthermore, we tested the methylation status of SST in biopsy samples, and the cell growth inhibitory effect of the SST analogue octreotide in human colorectal adenocarcinoma cell line. Colonic samples were collected from healthy children (n1 = 6), healthy adults (n2 = 41) and colorectal cancer patients (CRCs) (n3 = 34) for SST mRNA expression analysis, using HGU133 Plus2.0 microarrays. Results were validated both on original (n1 = 6; n2 = 6; n3 = 6) and independent samples ((n1 = 6; n2 = 6; n3 = 6) by real-time PCR. SST expressing cells were detected by immunohistochemistry on colonic biopsy samples (n1 = 14; n2 = 20; n3 = 23). The effect of octreotide on cell growth was tested on Caco-2 cell line. SST methylation percentage in biopsy samples (n1 = 5; n2 = 5; n3 = 9) was defined using methylation-sensitive restriction enzyme digestion. In case of normal aging SST mRNA expression did not alter, but decreased in cancer (p < 0.05). The ratio of SST immunoreactive cells was significantly higher in children (0.70% ± 0.79%) compared to CRC (0% ± 0%) (p < 0.05). Octreotide significantly increased the proportion of apoptotic Caco-2 cells. SST showed significantly higher methylation level in tumor samples (30.2% ± 11.6%) compared to healthy young individuals (3.5% ± 1.9%) (p < 0.05). In cancerous colonic mucosa the reduced SST production may contribute to the uncontrolled cell proliferation. Our observation that in colon cancer cells octreotide significantly enhanced cell death and attenuated cell proliferation suggests that SST may act as a regulator of epithelial cell kinetics. The inhibition of SST expression in CRC can be epigenetically regulated by promoter hypermethylation.
Sigma-Aldrich
Octreotide, ≥98% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%