Merck
  • Preparation of heparan sulfate-like polysaccharide and application in stem cell chondrogenic differentiation.
Preparation of heparan sulfate-like polysaccharide and application in stem cell chondrogenic differentiation.
Carbohydrate research (2014-12-03)
Shancheng Zhao, Zhen Wang, Jingxiao Chen, Jinghua Chen
ABSTRACT
Heparan sulfate is a component of the extracellular matrix (ECM) that modulates individual development and cell growth through its interaction with growth factors. Structurally, heparan sulfate consists of repeating linear sulfated poly-anionic disaccharide structures. The K5 polysaccharide has the same structure as heparosan, and is the capsular polysaccharide of Escherichia coli K5 strain which serves as a precursor in heparin and heparan sulfate biosynthesis. Here, we prepared sulfated K5 polysaccharides that are structurally similar to heparan sulfate and investigated their biocompatibility and bioactivity in stem cell chondrogenic differentiation. Briefly, sulfation groups were added to -NH- and/or -OH of a precursor heparosan and the modified heparosan was qualitatively analyzed by FT-IR, (1)H NMR, and (13)C NMR techniques. Cell viability was not significantly affected by the sulfated K5 capsular polysaccharide. Relative mRNA expression of the chondrogenic differentiation marker COL2A1 was significantly upregulated in cells treated with the N,O-sulfated K5 polysaccharide confirming that the sulfated K5 capsular polysaccharide is able to stimulate chondrogenic differentiation. The main sulfation pattern for chondrogenic activity is N,6-O sulfation and the activity was not proportional to the sulfation level. This type of mimic was prepared in nearly a gram scale, supporting further structural study and 3 dimension stem cell culture. Together, the results of this study show that sulfated K5 capsular polysaccharides are able to stimulate chondrogenic differentiation without affecting cell viability.
Product Description
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
Sulfur, PESTANAL®, analytical standard
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)