Merck
  • A microwave radiosynthesis of the 4-[18F]-fluorobenzyltriphenylphosphonium ion.
A microwave radiosynthesis of the 4-[18F]-fluorobenzyltriphenylphosphonium ion.
Journal of labelled compounds & radiopharmaceuticals (2014-10-21)
Hayden T Ravert, Daniel P Holt, Robert F Dannals
ABSTRACT
The 4-[(18)F]-fluorobenzyltriphenylphosphonium cation was synthesized by a series of microwave reactions from no carrier added [(18)F]-fluoride. The microwave procedure reduced the quantity of reagents used and synthesis time when compared with the original synthesis. In addition, problematic solid phase extraction, sodium borohydride reduction by column and inconsistent yields with excessive precipitate formation during the bromination step were eliminated. The 4-[(18)F]-fluorobenzyltriphenylphosphonium cation was produced radiochemically pure in 8.3% yield with a specific radioactivity of 534.5 ± 371.4 GBq/µmole at end of synthesis.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen bromide solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Hydrobromic acid solution, 33 wt. % in acetic acid