Merck
  • A novel E2F/sphingosine kinase 1 axis regulates anthracycline response in squamous cell carcinoma.
A novel E2F/sphingosine kinase 1 axis regulates anthracycline response in squamous cell carcinoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-11-21)
Mehlika Hazar-Rethinam, Lilia Merida de Long, Orla M Gannon, Eleni Topkas, Samuel Boros, Ana Cristina Vargas, Marcin Dzienis, Pamela Mukhopadhyay, Fiona Simpson, Liliana Endo-Munoz, Nicholas A Saunders
ABSTRACT
Head and neck squamous cell carcinomas (HNSCC) are frequently drug resistant and have a mortality rate of 45%. We have previously shown that E2F7 may contribute to drug resistance in SCC cells. However, the mechanism and pathways involved remain unknown. We used transcriptomic profiling to identify candidate pathways that may contribute to E2F7-dependent resistance to anthracyclines. We then manipulated the activity/expression of the candidate pathway using overexpression, knockdown, and pharmacological inhibitors in in vitro and in vivo models of SCC to demonstrate causality. In addition, we examined the expression of E2F7 and a downstream effector in a tissue microarray (TMA) generated from HNSCC patient samples. E2F7-deficient keratinocytes were selectively sensitive to doxorubicin and this was reversed by overexpressing E2F7. Transcriptomic profiling identified Sphingosine kinase 1 (Sphk1) as a potential mediator of E2F7-dependent drug resistance. Knockdown and overexpression studies revealed that Sphk1 was a downstream target of E2F7. TMA studies showed that E2F7 overexpression correlated with Sphk1 overexpression in human HNSCC. Moreover, inhibition of Sphk1 by shRNA or the Sphk1-specific inhibitor, SK1-I (BML-EI411), enhanced the sensitivity of SCC cells to doxorubicin in vitro and in vivo. Furthermore, E2F7-induced doxorubicin resistance was mediated via Sphk1-dependent activation of AKT in vitro and in vivo. We identify a novel drugable pathway in which E2F7 directly increases the transcription and activity of the Sphk1/S1P axis resulting in activation of AKT and subsequent drug resistance. Collectively, this novel combinatorial therapy can potentially be trialed in humans using existing agents.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
D-Sphingosine, synthetic
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy