Merck
  • Induction of apoptosis in human peritoneal mesothelial cells by gastric cancer cell supernatant promotes peritoneal carcinomatosis.
Induction of apoptosis in human peritoneal mesothelial cells by gastric cancer cell supernatant promotes peritoneal carcinomatosis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-05-27)
Di Na, Yan Song, Cheng-Gang Jiang, Zhe Sun, Ying-Ying Xu, Zhen-Ning Wang, Zhuo-Zheng Zhao, Hui-Mian Xu
ABSTRACT
This study was carried out to evaluate the effects of gastric cancer cell supernatant on human peritoneal mesothelial cell viability and apoptosis and to investigate the mechanism of action of gastric cancer in a mesothelial cell line (HMrSV5). Cell viability was measured using the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazoliumbromide (MTT) assay. Mesothelial cells treated with gastric cancer cell supernatant were stained with acridine orange/ethidium bromide and subjected to fluorescence microscopy. C57BL/6 mice were used to establish a cancer invasion model. Morphological changes and exfoliation occurred, and naked areas appeared in both cultured mesothelial cells and the parietal peritoneum after treatment with gastric cancer cell supernatant. Cell supernatant from gastric cancer cells induced apoptosis of mesothelial cells in a time-dependent manner. Obvious morphological changes of cell apoptosis were detected, such as condensation of chromatin, nuclear fragmentations, and apoptotic ladders. These findings demonstrate that gastric cancer cells induce apoptosis of human peritoneal mesothelial cells through supernatants in early peritoneal metastasis.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
5-Methylthiazole, 97%
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES