Merck
The protective effect of selenium in cisplatin-related retinotoxicity.
Cutaneous and ocular toxicology (2014-03-20)
Alper Yazici, Esin Sogutlu-Sari, Arzu Yay, Hasan Aksit, Adil Kilic, Dilek Aksit, Onur Yildiz, Sitki Samet Ermis
ABSTRACT
The aim of this study is to evaluate the retinal toxicity of cisplatin and neuroprotective effect of selenium in cisplatin-related retinal toxicity. Eighteen adult Wistar-Albino rats were divided into three groups. Group 1 (n = 6) received intraperitoneal (i.p.) injection of 2.5 ml physiologic saline for three days, group 2 (n = 6) received i.p. 16 mg/kg cisplatin for three days and group 3 (n = 6) received i.p. 16 mg/kg cisplatin for three days and 1.5 mg/kg twice daily selenium via gavage five days prior to cisplatin injection and for three days concomitantly with cisplatin injections. The total retinal thickness, outer nuclear layer (ONL), inner nuclear layer (INL) and inner plexiform layer (IPL) thicknesses were measured in hematoxylin/eosin and apoptotic index (AI) of ganglion cell layer (GCL) and INL was evaluated in TdT-mediated dUTP-biotin nick end labeling (TUNEL)-stained retina sections. Selenium statistically succeeded to reduce total retinal thickness in cisplatin-toxicated retinas: from 210.17 ± 23.40 to 173.55 ± 20.43, ONL: 49.79 ± 5.32 to 41.87 ± 6.30, INL: 33.72 ± 7.93 to 25.06 ± 5.73 and IPL: 53.61 ± 8.63 to 45.61 ± 6.92 µm in hematoxylin/eosin-stained retina sections. The AI was also reduced in INL (30.10 ± 12.02 to 19.48 ± 12.99) and in GCL (37.59 ± 17.70 to 33.15 ± 13.78). However, statistical significance was present in only AI values of INL. Selenium limited edema due to the toxicity and reduced the retinal thickness and showed neuroprotection in cisplatin-induced retinotoxicity.
Product Description
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline