Merck
  • PPM1D is a prognostic marker and therapeutic target in colorectal cancer.
PPM1D is a prognostic marker and therapeutic target in colorectal cancer.
Experimental and therapeutic medicine (2014-07-11)
Tian-Shu Peng, Yong-Heng He, Tian Nie, Xiang-Dang Hu, Hai-Yan Lu, Jian Yi, Yun-Fei Shuai, Min Luo
ABSTRACT
Protein phosphatase, Mg
Sigma-Aldrich
MISSION® esiRNA, targeting human PPM1D
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ppm1d