Merck
  • Mixed carbonates as useful substrates for a fluorogenic assay for lipases and esterases.
Mixed carbonates as useful substrates for a fluorogenic assay for lipases and esterases.
Chembiochem : a European journal of chemical biology (2015-02-05)
Anna Zadlo, Dominik Koszelewski, Filip Borys, Ryszard Ostaszewski
ABSTRACT
Mixed carbonates of 7-hydroxy-4-methylcoumarin were shown to be a new class of probe for fluorogenic assays. They are promising substrates for fingerprinting enzyme hydrolytic activity, and proved particularly useful because of the low level of nonspecific degradation and ease of synthesis. They are highly relevant for screening lipase and esterase libraries. These advantages make umbelliferyl carbonates highly suitable substrates for high-throughput screening. Moreover, we report the use of chiral fluorogenic carbonates as enantiopreference probes.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hexane, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)