Merck
  • Identification and characterization of functional Smad8 and Smad4 homologues from Echinococcus granulosus.
Identification and characterization of functional Smad8 and Smad4 homologues from Echinococcus granulosus.
Parasitology research (2014-07-21)
Chuanshan Zhang, Limin Wang, Hui Wang, Hongwei Pu, Le Yang, Jing Li, Junhua Wang, Guodong Lü, Xiaomei Lu, Wenbao Zhang, Dominique A Vuitton, Hao Wen, Renyong Lin
ABSTRACT
Smad family proteins are essential cellular mediators of the transforming growth factor-β superfamily. In the present study, we identified two members of the Smad proteins, Smad8 and Smad4 homologues (termed as EgSmadE and EgSmadD, respectively), from Echinococcus granulosus, the causative agent of cystic echinococcosis (CE). Phylogenetic analysis placed EgSmadE in the Smad1, 5, and 8 subgroup of the R-Smad sub-family and EgSmadD in the Co-Smad family. Furthermore, EgSmadE and EgSmadD attained a high homology to EmSmadE and EmSmadD of E. multilocularis, respectively. Both EgSmadE and EgSmadD were co-expressed in the larval stages and exhibited the highest transcript levels in activated protoscoleces, and their encoded proteins were co-localized in the sub-tegumental and tegumental layer of the parasite. As shown by yeast two-hybrid and pull-down analysis, EgSmadE displayed a positive binding interaction with EgSmadD. In addition, EgSmadE localized in the nuclei of Mv1Lu cells (mink lung epithelial cells) upon treatment with human TGF-β1 or human BMP2, indicating that EgSmadE is capable of being translocated into nucleus, in vitro. Our study suggests that EgSmadE and EgSmadD may take part in critical biological processes, including echinococcal growth, development, and parasite-host interaction.
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard