Merck
  • Protein kinase D2 induces invasion of pancreatic cancer cells by regulating matrix metalloproteinases.
Protein kinase D2 induces invasion of pancreatic cancer cells by regulating matrix metalloproteinases.
Molecular biology of the cell (2013-12-18)
Christoph Wille, Conny Köhler, Milena Armacki, Arsia Jamali, Ulrike Gössele, Klaus Pfizenmaier, Thomas Seufferlein, Tim Eiseler
ABSTRACT
Pancreatic cancer cell invasion, metastasis, and angiogenesis are major challenges for the development of novel therapeutic strategies. Protein kinase D (PKD) isoforms are involved in controlling tumor cell motility, angiogenesis, and metastasis. In particular PKD2 expression is up-regulated in pancreatic cancer, whereas PKD1 expression is lowered. We report that both kinases control pancreatic cancer cell invasive properties in an isoform-specific manner. PKD2 enhances invasion in three-dimensional extracellular matrix (3D-ECM) cultures by stimulating expression and secretion of matrix metalloproteinases 7 and 9 (MMP7/9), by which MMP7 is likely to act upstream of MMP9. Knockdown of MMP7/9 blocks PKD2-mediated invasion in 3D-ECM assays and in vivo using tumors growing on chorioallantois membranes. Furthermore, MMP9 enhances PKD2-mediated tumor angiogenesis by releasing extracellular matrix-bound vascular endothelial growth factor A, increasing its bioavailability and angiogenesis. Of interest, specific knockdown of PKD1 in PKD2-expressing pancreatic cancer cells further enhanced the invasive properties in 3D-ECM systems by generating a high-motility phenotype. Loss of PKD1 thus may be beneficial for tumor cells to enhance their matrix-invading abilities. In conclusion, we define for the first time PKD1 and 2 isoform-selective effects on pancreatic cancer cell invasion and angiogenesis, in vitro and in vivo, addressing PKD isoform specificity as a major factor for future therapeutic strategies.
Sigma-Aldrich
MISSION® esiRNA, targeting human PKD1
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKD1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Prkd1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pkd1