Merck
  • A TTX-sensitive resting Na+ permeability contributes to the catecholaminergic automatic activity in rat pulmonary vein.
A TTX-sensitive resting Na+ permeability contributes to the catecholaminergic automatic activity in rat pulmonary vein.
Journal of cardiovascular electrophysiology (2014-10-28)
Claire O Malécot, Pierre Bredeloux, Ian Findlay, Véronique Maupoil
ABSTRACT
Ectopic activity arising from pulmonary veins (PV) plays a prominent role in the onset of atrial fibrillation in humans. Rat PV cardiac muscle cells have a lower resting membrane potential (RMP) than the left atria (LA) and presents in the presence of norepinephrine an automatic activity, which occurs in bursts. This study investigated the role of Na channels upon the RMP and the catecholaminergic automatic activity (CAA) in PV cardiac muscle. RMP and CAA experiments were performed in male Wistar rat PV. Whole-cell INa was recorded in isolated PV and LA cardiomyocytes. PV has a higher tetrodotoxin (TTX)-sensitive basal Na(+) permeability than the LA, due to a ∼ 5 mV more negative Na window current in the former tissue. TTX, quinidine, and ranolazine (1 to 10 μM each) decreased CAA incidence and arrhythmias by increasing burst intervals because of a reduction of the slope of slow depolarization between bursts. TTX and ranolazine also reduced burst duration. At 1 Hz, 10 μM quinidine, ranolazine, and TTX inhibited peak INa by 33%, 28%, and 98%, respectively. Each reduced the Na window current. There was no evidence for a TTX- or ranolazine-sensitive late Na current. Na channels confer a TTX-sensitive basal Na(+) permeability to rat PV cardiac muscle cells and contribute to the slope of slow depolarization between bursts of CAA. Na channel blockers act mostly via reduction of the Na window current. Ranolazine also has an anti-α1 adrenergic effect, which contributed to its antiarrhythmic effect.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%