Merck
  • Stereoselective recognition of amethopterin enantiomers by the rat proton-coupled folate transporter.
Stereoselective recognition of amethopterin enantiomers by the rat proton-coupled folate transporter.
Biological & pharmaceutical bulletin (2015-04-04)
Tomoya Narawa, Toshiaki Yano, Tomoo Itoh
ABSTRACT
The stereoselective transport of methotrexate (L-amethopterin, L-MTX) and its enantiomer (D-amethopterin, D-MTX) by the rat proton-coupled folate transporter (rPCFT) were examined using rPCFT-expressing HEK293 cells. The initial rate of uptake of [3H]-L-MTX by the rPCFT followed Michaelis-Menten kinetics, with a Km value of 2.1 µM. Dixon plots revealed that the uptake of L-MTX by the rPCFT was inhibited in a competitive manner by unlabeled L-MTX and D-MTX, with Ki values of approximately 1.3 and 150 µM, respectively. The initial rate of uptake of D-MTX by the rPCFT also followed Michaelis-Menten kinetics, with a Km value of 190 µM. The results of the current study demonstrate that the different enantiomers of MTX are transported in a highly stereoselective manner by the rPCFT, with the uptake clearance of L-MTX being approximately 46-fold greater than that of D-MTX. The observed stereoselectivity of the rPCFT was found to be comparable with that of the human PCFT.
Sigma-Aldrich
MES monohydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
G 418 disulfate salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
G 418 disulfate salt, potency: ≥720 μg per mg (dried basis)
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES monohydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
SAFC
HEPES
SAFC
MES monohydrate
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MES solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate