Merck
  • Stabilities of neutral and basic esters of bendamustine in plasma compared to the parent compound: kinetic investigations by HPLC.
Stabilities of neutral and basic esters of bendamustine in plasma compared to the parent compound: kinetic investigations by HPLC.
Journal of pharmaceutical and biomedical analysis (2014-12-17)
S Huber, F Antoni, C Schickaneder, H Schickaneder, G Bernhardt, A Buschauer
ABSTRACT
Esters of the cytostatic bendamustine (1), previously demonstrated to be much more potent than the parent compound as antiproliferative agents in vitro, were investigated for stability in buffer and plasma, as well as against porcine liver esterase in the presence of different amounts of albumin using a validated RP-HPLC method with fluorescence detection. The hydrolysis of the nitrogen mustard moiety was retarded (for 1: approximately 130 vs. 11 min) in the presence of plasma proteins. For the derivatives, both cleavage of ester and nitrogen mustard moieties were analyzed. Enzymatic hydrolysis was very fast in the case of 2-pyrrolidino-, 2-piperidino- and 2-(4-methylpiperazino)-ethyl esters, whereas methyl, ethyl, morpholinoethyl and branched 2-pyrrolidinoethyl esters were considerably more stable (half-lives between 41 and 116 min, compared to <5 min). Inhibition by physostigmine indicated unspecific cholinesterases to be involved in the rapid ester cleavage. Due to lower protein content and higher enzymatic activity in murine compared to human plasma, reduced stability of all investigated esters in mouse plasma (t½<2 min) has to be taken into account with respect to the design of animal studies.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Umbelliferone, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Umbelliferone, analytical standard
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material