Merck
  • Assay of total glutathione and glutathione disulphide in seminal plasma of male partners of couples presenting for a fertility evaluation.
Assay of total glutathione and glutathione disulphide in seminal plasma of male partners of couples presenting for a fertility evaluation.
Andrologia (2013-10-16)
R Kanďár, N Hájková
ABSTRACT
A method is described here for the determination of total glutathione (TGSH) and glutathione disulphide (GSSG) in the seminal plasma of the male partners of couples requesting a fertility evaluation. A suitable sample preparation procedure prior to high-performance liquid chromatography analysis is discussed. After adequate sample preparation, the samples were derivatised with ortho-phthaldialdehyde to form a stable, highly fluorescent tricyclic derivative. Reversed-phase column chromatography was used for the separation, and the effluent was monitored with a fluorescence detector at an excitation wavelength of 350 nm and an emission wavelength of 420 nm. The analytical performance of this method was satisfactory. The intra-assay and inter-assay coefficients of variation were below 10%. The recoveries were as follows: 94.1% (CV 2.3%) for TGSH and 93.2% (CV 4.0%) for GSSG. No significant differences were found in either TGSH or GSSG concentration between the smokers and nonsmokers (2.07 ± 1.28 μm versus 1.56 ± 1.20 μm, P = 0.431 and 95 ± 56 nm versus 112 ± 138 nm, P = 0.825).
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard