Merck
  • Polybrominated diphenyl ethers in indoor air during waste TV recycling process.
Polybrominated diphenyl ethers in indoor air during waste TV recycling process.
Journal of hazardous materials (2014-12-03)
Jie Guo, Kuangfei Lin, Jingjing Deng, Xiaoxu Fu, Zhenming Xu
ABSTRACT
Recycling process for waste TV sets mainly consists of dismantling, printed wiring board (PWB) heating, PWB recycling, and plastic crushing in formal recycling plant. Polybrominated diphenyl ethers (PBDEs) contained in waste TV sets are released to indoor air. Air samples at 4 different workshops were collected to measure the PBDEs concentrations in both gaseous and particulate phases. The mean concentrations of ∑PBDEs in indoor air were in the range of 6780-2,280,000 pg/m(3). The highest concentration in gaseous phase (291,000 pg/m(3)) was detected in the PWB heating workshop. The ∑12PBDEs concentrations in PM2.5 and PM10 at the 4 workshops ranged in 6.8-6670 μg/g and 32.6-6790 μg/g, respectively. The gas-particle partitioning of PBDEs was disrupted as PBDEs were continuously released during the recycling processes. Occupational exposure assessment showed that only the exposure concentration of BDE-47 (0.118 μg/kg/day) through inhalation in the PWB heating workshop for workers without facemask exceeded the reference dose (0.1 μg/kg/day), posing a health hazard to workers. All the results demonstrated that recycling of TV sets was an important source of PBDEs emission, and PBDEs emission pollution was related to the composition of TV sets, interior dust, and recycling process.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer