Merck
  • Development of a pharmacokinetic/pharmacodynamic/disease progression model in NC/Nga mice for development of novel anti-atopic dermatitis drugs.
Development of a pharmacokinetic/pharmacodynamic/disease progression model in NC/Nga mice for development of novel anti-atopic dermatitis drugs.
Xenobiotica; the fate of foreign compounds in biological systems (2014-05-31)
In-Hwan Baek, Byung-Yo Lee, Jung-Woo Chae, Gyu Yong Song, Wonku Kang, Kwang-Il Kwon
ABSTRACT
1. JHL45, a novel immune modulator against atopic dermatitis (AD), was synthesized from decursin isolated from Angelica gigas. The goal is to evaluate the lead compound using quantitative modeling approaches to novel anti-AD drug development. 2. We tested the anti-inflammatory effect of JHL45 by in vitro screening, characterized its in vitro pharmacokinetic (PK) properties. The dose-dependent efficacy of JHL45 was developed using a pharmacokinetics/pharmacodynamics/disease progression (PK/PD/DIS) model in NC/Nga mice. 3. JHL45 has drug-like properties and pharmacological effects when administered orally to treat atopic dermatitis. The developed PK/PD/DIS model described well the rapid metabolism of JHL45, double-peak phenomenon in the PK of decursinol and inhibition of IgE generation by compounds in NC/Nga mice. Also, a quantitative model was developed and used to elucidate the complex interactions between serum IgE concentration and atopic dermatitis symptoms. 4. Our findings indicate that JHL45 has good physicochemical properties and powerful pharmacological effects when administered orally for treatment of AD in rodents.
Product Description
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Supelco
Umbelliferone, analytical standard
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)