Merck
  • Purification and characterization of parvalbumin isotypes from grass carp (Ctenopharyngodon idella).
Purification and characterization of parvalbumin isotypes from grass carp (Ctenopharyngodon idella).
Journal of agricultural and food chemistry (2014-05-29)
Zheng Li, Juan You, Yongkang Luo, Jianping Wu
ABSTRACT
The prevalence of fish allergy is rapidly increasing because of a growing fish consumption driven mainly by a positive image of the fish and health relationship. The purpose of this study was to characterize parvalbumin isotypes from grass carp (Ctenopharyngodon idella), one of the most frequently consumed freshwater fish in China. Three parvalbumin isotypes were purified using consecutive gel filtration and reverse-phase chromatography and denoted as PVI, PVII, and PVIII. The molecular weights of the isotypes were determined to be 11.968, 11.430, and 11.512 kDa, respectively. PVI showed 74% matched amino acids sequence with PV isotype 4a from Danio rerio, while PVII and PVIII showed 46% matched amino acids sequence with PV isotypes from Hypophthalmichthys molitrix. PVII is the dominant allergen, but it was liable to gastrointestinal enzymes as PVIII; however, PVI was resistant to pepsin digestion. A further study is to characterize the epitopes of PVII, the dominant allergen.
Product Description
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis