Merck
  • The efflux pump inhibitor timcodar improves the potency of antimycobacterial agents.
The efflux pump inhibitor timcodar improves the potency of antimycobacterial agents.
Antimicrobial agents and chemotherapy (2014-12-24)
Trudy H Grossman, Carolyn M Shoen, Steven M Jones, Peter L Jones, Michael H Cynamon, Christopher P Locher
ABSTRACT
Previous studies indicated that inhibition of efflux pumps augments tuberculosis therapy. In this study, we used timcodar (formerly VX-853) to determine if this efflux pump inhibitor could increase the potency of antituberculosis (anti-TB) drugs against Mycobacterium tuberculosis in in vitro and in vivo combination studies. When used alone, timcodar weakly inhibited M. tuberculosis growth in broth culture (MIC, 19 μg/ml); however, it demonstrated synergism in drug combination studies with rifampin, bedaquiline, and clofazimine but not with other anti-TB agents. When M. tuberculosis was cultured in host macrophage cells, timcodar had about a 10-fold increase (50% inhibitory concentration, 1.9 μg/ml) in the growth inhibition of M. tuberculosis and demonstrated synergy with rifampin, moxifloxacin, and bedaquiline. In a mouse model of tuberculosis lung infection, timcodar potentiated the efficacies of rifampin and isoniazid, conferring 1.0 and 0.4 log10 reductions in bacterial burden in lung, respectively, compared to the efficacy of each drug alone. Furthermore, timcodar reduced the likelihood of a relapse infection when evaluated in a mouse model of long-term, chronic infection with treatment with a combination of rifampin, isoniazid, and timcodar. Although timcodar had no effect on the pharmacokinetics of rifampin in plasma and lung, it did increase the plasma exposure of bedaquiline. These data suggest that the antimycobacterial drug-potentiating activity of timcodar is complex and drug dependent and involves both bacterial and host-targeted mechanisms. Further study of the improvement of the potency of antimycobacterial drugs and drug candidates when used in combination with timcodar is warranted.
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Phenol Red, ACS reagent
Ethionamide for system suitability, European Pharmacopoeia (EP) Reference Standard
Clofazimine, European Pharmacopoeia (EP) Reference Standard
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethidium bromide, BioReagent, powder
Sigma-Aldrich
Clofazimine
Aucubin, primary reference standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Acriflavine, fluorescent label
Sigma-Aldrich
Ethionamide
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Supelco
Aucubin, analytical standard
Sigma-Aldrich
Ethanolamine, ≥98%
Ethionamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Acriflavine, BioReagent, suitable for fluorescence, ≥90% (AT)
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, 10 mg/mL in H2O
Sigma-Aldrich
Ethanolamine, ≥99%