Merck
  • Human hepatitis B virus surface and e antigens inhibit major vault protein signaling in interferon induction pathways.
Human hepatitis B virus surface and e antigens inhibit major vault protein signaling in interferon induction pathways.
Journal of hepatology (2014-12-08)
Shi Liu, Nanfang Peng, Jiajia Xie, Qian Hao, Mo Zhang, Yi Zhang, Zhangchuan Xia, Gang Xu, Fanpeng Zhao, Qing Wang, Tao Han, Ying Zhu
ABSTRACT
We previously demonstrated that major vault protein (MVP) is a novel virus-induced host factor and its expression upregulates type-I interferon production, leading to cellular antiviral response. However, it remains unclear whether the antiviral function of MVP is impaired during hepatitis B virus (HBV) infection and what mechanisms are involved. Therefore, the aim of this study was to assess whether HBV can alter MVP expression despite the lack of type-I IFN induction and shed light on the underlying mechanisms HBV utilizes to evade host innate immune response. The ability of HBV surface and e antigens to inhibit MVP signaling in interferon induction pathways was evaluated by co-immunoprecipitation, immunofluorescence, quantitative RT-PCR, Western blot and reporter assays. In our current study, we found high levels of MVP in peripheral blood mononuclear cells, sera, and liver tissue from HBV-infected patients relative to healthy individuals. We determined that MVP intracellularly associates with MyD88, an adapter protein involved in virus-triggered induction of type-I IFN. Protein truncation analysis revealed that the middle domain of MVP (amino acid residues 310-620) was essential for MyD88 binding. Conversely, HBV inhibited MVP-induced type-I IFN production by suppressing MVP/MyD88 interaction. HBV antigens, both HBsAg and HBeAg, suppressed this interaction by competitively binding to the essential MyD88 binding region of MVP and limiting downstream IFN signaling. MVP is a virus-induced protein capable of binding with MyD88 leading to type-I IFN production. HBV may evade an immune response by disrupting this interaction and limiting type-I IFN antiviral activity.
Sigma-Aldrich
MISSION® esiRNA, targeting human MVP
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mvp