Merck
  • Characterization of Acinetobacter baumannii clinical isolates carrying bla(OXA-23) carbapenemase and 16S rRNA methylase armA genes in Yemen.
Characterization of Acinetobacter baumannii clinical isolates carrying bla(OXA-23) carbapenemase and 16S rRNA methylase armA genes in Yemen.
Microbial drug resistance (Larchmont, N.Y.) (2014-06-06)
Sofiane Bakour, Samer Ahmed Alsharapy, Abdelaziz Touati, Jean-Marc Rolain
ABSTRACT
The aim of this study was to investigate the molecular support of resistance to carbapenems, aminoglycosides, and fluoroquinolones in Acinetobacter baumannii clinical isolates collected from Yemen hospital. Three A. baumannii were isolated in February 2013 from three patients hospitalized at Al-Thawra University Hospital in Sana'a, Yemen. Antibiotic susceptibility testing was performed using the disk diffusion and E-test methods. Carbapenemase production was carried out by the modified Hodge test (MHT) and imipenem-ethylenediaminetetraacetic acid (EDTA) methods. Carbapenem, aminoglycoside, and fluoroquinolone resistance determinants were studied by polymerase chain reaction and sequencing. The epidemiological relatedness of the three strains was studied using multilocus sequence typing (MLST). The isolates were resistant to almost all antibiotics tested with very high imipenem, amikacin, and ciprofloxacin minimum inhibitory concentrations (>32, >256, and >32 mg/L, respectively). The microbiological tests showed that the three A. baumannii were MHT positive, besides, the activity of β-lactamases was not inhibited by EDTA. All the three isolates contained the naturally occurring bla(OXA-51)-like gene and the bla(OXA-23)-like carbapenemase-encoding gene. The 16S rRNA methylase armA gene was detected in the three isolates. In addition, screening for genes encoding the aminoglycoside-modifying enzymes (AMEs) demonstrated that one isolate contained the acetyltransferase gene aac(6')-Ib. Fluoroquinolone resistance was associated with a single mutation Ser83Leu in the quinolone resistance determining region of the gyrA gene in all isolates. The MLST showed that the sequence type (ST) obtained corresponds to ST2 for the three strains. Here we report the first identification of multidrug-resistant A. baumannii isolates harboring the bla(OXA-23)-like gene, AMEs [aac(6')-Ib], and the 16S rRNA methylase (armA) in the Yemen hospital.
Product Description
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material
Ceftazidime, European Pharmacopoeia (EP) Reference Standard
Millipore
Ticarcillin Supplement, suitable for microbiology
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Supelco
Aztreonam, analytical standard
USP
Tobramycin, United States Pharmacopeia (USP) Reference Standard
Tobramycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
USP
Amikacin, United States Pharmacopeia (USP) Reference Standard
Amikacin, European Pharmacopoeia (EP) Reference Standard
USP
Piperacillin, United States Pharmacopeia (USP) Reference Standard
Piperacillin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard