Merck
  • Synthesis and characterization of neutral and anionic cellulosic amphiphiles.
Synthesis and characterization of neutral and anionic cellulosic amphiphiles.
Carbohydrate polymers (2014-09-27)
Haoyu Liu, Kevin J Edgar
ABSTRACT
Polymer design and selection are crucial to the development of amorphous solid dispersions (ASD) for solubilization of otherwise poorly water-soluble drugs. The matrix polymer is required to interact strongly at the molecular level with the drug to prevent recrystallization, but must also be able to release the drug at an adequate rate upon entering the absorptive portion of the digestive tract. Herein we report versatile syntheses of a non-ionic, water-soluble cellulosic polymer family, cellulose trioxodecanoates, containing a hydrophilic oligo(ethylene oxide) side chain. This series of cellulose derivatives is designed for both adequate stabilization of amorphous drugs with high crystallization tendency, and timely release of those drugs. Alternatively, these polymers can be rendered anionic by also appending a pH-responsive ω-carboxyalkanoate group. Detailed structural information and structure-property relationship characterization of these amphiphilic polymers are described, which will permit evaluation of these materials as ASD polymers for enhancement of drug solubility and bioavailability.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Propionic anhydride, purum, ≥96.0% (NT)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Propionic anhydride, 97%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%
Sigma-Aldrich
Oxalyl chloride solution, 2.0 M in methylene chloride
Sigma-Aldrich
Oxalyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Propionic anhydride, ≥99%
Sigma-Aldrich
Oxalyl chloride, reagent grade, 98%
Sigma-Aldrich
CDI, reagent grade
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%