Merck
  • Induction of long-term immunity against respiratory syncytial virus glycoprotein by an osmotic polymeric nanocarrier.
Induction of long-term immunity against respiratory syncytial virus glycoprotein by an osmotic polymeric nanocarrier.
Acta biomaterialia (2014-08-12)
Jannatul Firdous, Mohammad Ariful Islam, Sung-Moo Park, In-Su Cheon, Byoung-Shik Shim, Hyo-Shin Yoon, Manki Song, Jun Chang, Yun-Jaie Choi, Yeong-Min Park, Diana Boraschi, Seung-Hyun Han, Chong-Su Cho, Cheol-Heui Yun
ABSTRACT
Respiratory syncytial virus (RSV) is one of the most common causes of viral deaths in infants worldwide, yet no effective vaccines are available. Here, we report an osmotically active polysaccharide-based polysorbitol transporter (PST) prepared from sorbitol diacrylate and low-molecular-weight polyethylenimine (PEI) showing a potent, yet safe, adjuvant activity and acting as an effective delivery tool for RSV glycoprotein (RGp) antigen. PST showed no toxicity in vitro or in vivo, unlike PEI and the well-known experimental mucosal adjuvant cholera toxin (CT). PST formed nano-sized complexes with RGp by simple mixing, without affecting antigenic stability. The complexes exhibited negative surface charges that made them highly efficient in the selective activation of phagocytic cells and enhancement of phagocytic uptake. This resulted in an improved cytokine production and in the significant augmentation of RGp-specific antibody production, which persisted for over 200 days. Interestingly, PST/RGp enhanced phagocytic uptake owing to the osmotic property of PST and its negative zeta potential, suggesting that PST could selectively stimulate phagocytic cells, thereby facilitating a long-lived antigen-specific immune response, which was presumably further enhanced by the polysaccharide properties of PST.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)