Merck
  • Synthesis of azobenzenealkylmaleimide probes to photocontrol the enzyme activity of a bacterial histone deacetylase-like amidohydrolase.
Synthesis of azobenzenealkylmaleimide probes to photocontrol the enzyme activity of a bacterial histone deacetylase-like amidohydrolase.
Bioorganic chemistry (2014-12-03)
Benjamin Horstmann, Michael Korbus, Tatjana Friedmann, Christiane Wolff, Christina Marie Thiele, Franz-Josef Meyer-Almes
ABSTRACT
A series of azobenzenealkylmaleimides (AMDs) with different spacer length was synthesized and coupled via Michael-Addition to a specific mutant of a bacterial histone deacetylase-like amidohydrolase (HDAH). Michaelis-Menten parameters (Vmax and Km) were employed to characterize the effect of both, the spacer length and the configuration (cis vs. trans) of the attached azobenzene moiety, on the HDAH enzyme activity. The photoswitch behavior of the AMD/enzyme conjugate activity was clearly influenced by the AMD spacer length. This study highlights the importance of steric rearrangement of the photoswitch with respect to the active site and describes a strategy to optimize the photocontrol of HDAH.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications