Merck
  • Anti-epidermal growth factor receptor conjugated mesoporous zinc oxide nanofibers for breast cancer diagnostics.
Anti-epidermal growth factor receptor conjugated mesoporous zinc oxide nanofibers for breast cancer diagnostics.
Nanoscale (2015-03-27)
Md Azahar Ali, Kunal Mondal, Chandan Singh, Bansi Dhar Malhotra, Ashutosh Sharma
ABSTRACT
We report the fabrication of an efficient, label-free, selective and highly reproducible immunosensor with unprecedented sensitivity (femto-molar) to detect a breast cancer biomarker for early diagnostics. Mesoporous zinc oxide nanofibers (ZnOnFs) are synthesized by electrospinning technique with a fiber diameter in the range of 50-150 nm. Fragments of ZnOnFs are electrophoretically deposited on an indium tin oxide glass substrate and conjugated via covalent or electrostatic interactions with a biomarker (anti-ErbB2; epidermal growth factor receptor 2). Oxygen plasma treatment of the carbon doped ZnOnFs generates functional groups (-COOH, -OH, etc.) that are effective for the conjugation of anti-ErbB2. ZnOnFs without plasma treatment that conjugate via electrostatic interactions were also tested for comparison. Label-free detection of the breast cancer biomarker by this point-of-care device is achieved by an electrochemical impedance technique that has high sensitivity (7.76 kΩ μM(-1)) and can detect 1 fM (4.34 × 10(-5) ng mL(-1)) concentration. The excellent impedimetric response of this immunosensor provides a fast detection (128 s) in a wide detection test range (1.0 fM-0.5 μM). The oxy-plasma treated ZnOnF immunoelectrode shows a higher association constant (404.8 kM(-1) s(-1)) indicating a higher affinity towards the ErbB2 antigen compared to the untreated ZnOnF immunoelectrode (165.6 kM(-1) s(-1)). This sensor is about an order of magnitude more sensitive than the best demonstrated in the literature based on different nanomaterials and about three orders of magnitude better than the ELISA standard for breast cancer biomarker detection. This proposed point-of-care cancer diagnostic offers several advantages, such as higher stability, rapid monitoring, simplicity, cost-effectiveness, etc., and should prove to be useful for the detection of other bio- and cancer markers.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Supelco
Naphthalene, analytical standard
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Supelco
Glycerol, analytical standard
Supelco
Melting point standard 79-81°C, analytical standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)