Merck
  • Quantitation of pregabalin in dried blood spots and dried plasma spots by validated LC-MS/MS methods.
Quantitation of pregabalin in dried blood spots and dried plasma spots by validated LC-MS/MS methods.
Journal of pharmaceutical and biomedical analysis (2015-03-15)
Nađa Kostić, Yannis Dotsikas, Nebojša Jović, Galina Stevanović, Anđelija Malenović, Mirjana Medenica
ABSTRACT
In this paper, novel LC-MS/MS methods for the determination of antiepileptic drug pregabalin in dried matrix spots (DMS) are presented. This attractive technique of sample collection in micro amount was utilized in the form of dried blood spots (DBS) and dried plasma spots (DPS). Following a pre-column derivatization procedure, using n-propyl chloroformate in the presence of n-propanol, and consecutive liquid-liquid extraction, derivatized pregabalin and its internal standard, 4-aminocyclohexanecarboxylic acid, were detected in positive ion mode by applying two SRM transitions per analyte. A YMC-Pack Octyl column (50mm×4.0mm, 3μm particle size) maintained at 30°C, was utilized with running mobile phase composed of acetonitrile: 0.15% formic acid (85:15, v/v). Flow rate was 550μL/min and total run time 2min. Established methods were fully validated over the concentration range of 0.200-20.0μg/mL for DBS and 0.400-40.0μg/mL for DPS, respectively, while specificity, accuracy, precision, recovery, matrix-effect, stability, dilution integrity and spot homogeneity were found within acceptance criteria. Validated methods were applied for the determination of pregabalin levels in dried blood and plasma samples obtained from patients with epilepsy, after per os administration of commercial capsules. Comparison of drug level in blood and plasma, as well as correction steps undertaken in order to overcome hematocrit issue, when analyzing DBS, are also given.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Chloroform, analytical standard
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material