Merck
  • Antimicrobial properties of nano-silver: a cautionary approach to ionic interference.
Antimicrobial properties of nano-silver: a cautionary approach to ionic interference.
Journal of colloid and interface science (2014-12-23)
K Sheehy, A Casey, A Murphy, G Chambers
ABSTRACT
Metallic nanoparticles such as nano-silver have found many applications as alternative antimicrobials in recent years. However methods for determining their proposed antimicrobial activity have received little attention to date. The disk diffusion assay is commonly used as a demonstration of antimicrobial properties and is a regular feature in synthetic nanoparticle papers. The aim of this study was to assess its effectiveness in demonstrating the "nanoparticle specific" antimicrobial properties in the absence of ionic contributions from unreacted reducing agents and or impurities. The disk diffusion assay was carried out on a range of silver nanoparticles, both in-house synthesised and commercially available, using Escherichia coli ATCC 25922 as a model organism. Capped and purified nanoparticles show no antimicrobial activity despite claims to the contrary for this assay. Results will be discussed in terms of the need for researchers without a background in microbiology to understand the mechanism of antimicrobial action before choosing an assay. Also discussed is the importance understanding the physiochemical characteristics of when interpreting results. Finally the relevance of the results in terms establishing protocols for method development for 'nanoparticle specific' antimicrobial properties will also be considered.
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%