Merck
  • Proteinase-activated receptor 1- and 4-promoted migration of Hep3B hepatocellular carcinoma cells depends on ROS formation and RTK transactivation.
Proteinase-activated receptor 1- and 4-promoted migration of Hep3B hepatocellular carcinoma cells depends on ROS formation and RTK transactivation.
Journal of cancer research and clinical oncology (2014-11-07)
Franziska Mußbach, Petra Henklein, Martin Westermann, Utz Settmacher, Frank-D Böhmer, Roland Kaufmann
ABSTRACT
There is growing evidence for a role of proteinase-activated receptors (PARs), a subfamily of G protein-coupled receptors, in cancer. We have previously shown that PAR1 and PAR4 are able to promote the migration of hepatocellular carcinoma (HCC) cells suggesting a function in HCC progression. In this study, we assessed the underlying signalling mechanisms. Using Hep3B liver carcinoma cells, RTK activation was assessed by Western blot employing phospho-RTK specific antibodies, ROS level were estimated by H2DCF-DA using confocal laser scanning microscopy, and measurement of PTP activity was performed in cell lysates using 6,8-difluoro-4-methylumbelliferyl phosphate (DiFMUP) as a substrate. Thrombin, the PAR1 selective agonist peptide TFLLRN-NH2 (PAR1-AP), and the PAR4 selective agonist peptide, AYPGKF-NH2 (PAR4-AP), induced a significant increase in Hep3B cell migration that could be blocked by inhibitors targeting formation of reactive oxygen species (ROS), or activation of hepatocyte-growth factor receptor (Met), or platelet-derived growth factor receptor (PDGFR), respectively. The involvement of these intracellular effectors in PAR1/4-initiated migratory signalling was further supported by the findings that individual stimulation of Hep3B cells with the PAR1-AP and the PAR4-AP induced an increase in ROS production and the transactivation of Met and PDGFR. In addition, PAR1- and PAR4-mediated inhibition of total PTP activity and specifically PTP1B. ROS inhibition by N-acetyl-L-cysteine prevented the inhibition of PTP1B phosphatase activity induced by PAR1-AP and the PAR4-AP, but had no effect on PAR1/4-mediated activation of Met and PDGFR in Hep3B cells. Collectively, our data indicate that PAR1 and PAR4 activate common promigratory signalling pathways in Hep3B liver carcinoma cells including activation of the receptor tyrosine kinases Met and PDGFR, the formation of ROS and the inactivation of PTP1B. However, PAR1/4-triggered Met and PDGFR transactivation seem to be mediated independently from the ROS-PTP1B signalling module.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
p-Terphenyl, ≥99.5% (HPLC)
Supelco
p-Terphenyl, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Malachite Green chloride, analytical standard