Merck
  • Sorting Nexin 27 regulates basal and activity-dependent trafficking of AMPARs.
Sorting Nexin 27 regulates basal and activity-dependent trafficking of AMPARs.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-30)
Natasha K Hussain, Graham H Diering, Jonathan Sole, Victor Anggono, Richard L Huganir
ABSTRACT
Activity-dependent changes in synaptic strength have long been postulated as cellular correlates of learning and memory. Long-term potentiation (LTP), a well characterized form of synaptic plasticity, is often expressed as an increase in the number of postsynaptic AMPA-type glutamate receptors (AMPARs). Although the precise molecular mechanisms governing LTP remain elusive, this study identifies one member of the sorting nexin family, Sorting Nexin 27 (SNX27), as a critical component in this process. The ability of sorting nexins to bind specific phospholipids as well as their propensity to form protein-protein complexes, points to a role for these proteins in membrane trafficking and protein sorting. Here, we demonstrate that SNX27 binds to AMPARs, and that this interaction is regulated in an activity-dependent manner. Furthermore, we provide evidence that SNX27 is synaptically enriched and its level of expression regulates targeting of AMPARs to the neuronal surface. Loss of SNX27 abolishes recruitment of surface AMPARs during chemical LTP. Collectively, our data suggest a role for SNX27 in modulating synaptic plasticity through regulated interaction with AMPARs.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material